Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112986.1 | NP_001106457.1 | 7 - 8 | 919 | 101 | |
GenBank mRNA | BC154067.1 | - | 7 - 8 | 919 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |