Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enosf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113128.1 | NP_001106599.1 | 9 - 10 | 625 | 101 | |
XM_012964234.2 | XP_012819688.1 | 10 - 11 | 1055 | 101 | ||
XM_012964235.2 | XP_012819689.1 | 10 - 11 | 972 | 101 | ||
XM_012964236.2 | XP_012819690.1 | 10 - 11 | 946 | 101 | ||
XM_018094523.1 | XP_017950012.1 | 9 - 10 | 1144 | 101 | ||
GenBank mRNA | BC155430.1 | - | 9 - 10 | 625 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |