Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113812.1 | NP_001107284.1 | 2 - 3 | 243 | 89 | |
GenBank mRNA | BC155537.1 | - | 2 - 3 | 243 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |