Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113890.1 | NP_001107362.1 | 5 - 6 | 686 | 77 | |
XM_012960563.2 | XP_012816017.1 | 6 - 7 | 1232 | 77 | ||
XM_012960564.2 | XP_012816018.1 | 6 - 7 | 1280 | 77 | ||
XM_012960565.1 | XP_012816019.1 | 6 - 7 | 753 | 77 | ||
GenBank mRNA | BC082746.1 | - | 6 - 7 | 755 | 77 | |
BC157213.1 | - | 5 - 6 | 686 | 77 | ||
BC171074.1 | - | 5 - 6 | 579 | 77 | ||
BC171076.1 | - | 5 - 6 | 579 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |