Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113906.1 | NP_001107378.1 | 2 - 3 | 727 | 100 | |
GenBank mRNA | BC157250.1 | - | 2 - 3 | 727 | 100 | |
BC170962.1 | - | 2 - 3 | 424 | 100 | ||
BC170964.1 | - | 2 - 3 | 424 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |