Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114031.1 | NP_001107503.1 | 6 - 7 | 700 | 67 | |
XM_012964257.2 | XP_012819711.1 | 751 | 67 | |||
GenBank mRNA | BC157661.1 | - | 6 - 7 | 700 | 67 | |
CR926350.2 | - | 659 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |