Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114063.2 | NP_001107535.2 | 6 - 7 | 677 | 110 | |
GenBank mRNA | CX742176.1 | - | 6 - 7 | 605 | 110 | |
EL854956.1 | - | 5 - 6 | 456 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |