Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114223.1 | NP_001107695.1 | 6 - 7 | 759 | 61 | |
XM_012968586.2 | XP_012824040.1 | 7 - 8 | 865 | 61 | ||
GenBank mRNA | BC121605.1 | - | 5 - 6 | 589 | 61 | |
BC155376.1 | - | 6 - 7 | 759 | 61 | ||
BC160442.1 | - | 7 - 8 | 832 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |