Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114489.1 | NP_001107961.1 | 5 - 6 | 810 | 86 | |
XM_012971718.2 | XP_012827172.1 | 6 - 7 | 933 | 86 | ||
XM_018097721.1 | XP_017953210.1 | 4 - 5 | 767 | 86 | ||
GenBank mRNA | BC088607.1 | - | 5 - 6 | 647 | 86 | |
BC118711.1 | - | 5 - 6 | 787 | 86 | ||
BC159128.1 | - | 5 - 6 | 810 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |