Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123413.1 | NP_001116885.1 | 5 - 6 | 1004 | 84 | |
GenBank mRNA | BC159140.1 | - | 5 - 6 | 1004 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |