Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123447.1 | NP_001116919.1 | 5 - 6 | 556 | 76 | |
XM_012960584.2 | XP_012816038.1 | 6 - 7 | 668 | 76 | ||
XM_012960585.2 | XP_012816039.1 | 6 - 7 | 752 | 76 | ||
XM_018092916.1 | XP_017948405.1 | 7 - 8 | 775 | 76 | ||
GenBank mRNA | BC161493.1 | - | 5 - 6 | 556 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |