Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lcmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126499.1 | NP_001119971.1 | 1707 | 96 | ||
GenBank mRNA | BC158186.1 | - | 1707 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |