Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126658.1 | NP_001120130.1 | 7 - 8 | 1128 | 91 | |
GenBank mRNA | BC159054.1 | - | 7 - 8 | 1128 | 91 | |
CT030321.1 | - | 7 - 8 | 1228 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |