Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpzl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126663.1 | NP_001120135.1 | 5 - 6 | 742 | 70 | |
GenBank mRNA | BC159074.1 | - | 5 - 6 | 742 | 70 | |
CU075738.1 | - | 5 - 6 | 705 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |