Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126812.1 | NP_001120284.1 | 2 - 3 | 1668 | 74 | |
GenBank mRNA | BC160551.1 | - | 2 - 3 | 1668 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |