Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: creb3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126879.1 | NP_001120351.1 | 815 | 75 | ||
GenBank mRNA | BC160986.1 | - | 815 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |