Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126942.1 | NP_001120414.1 | 3 - 4 | 322 | 75 | |
XM_012970087.2 | XP_012825541.1 | 3 - 4 | 2215 | 75 | ||
XM_012970089.2 | XP_012825543.1 | 3 - 4 | 288 | 75 | ||
XM_012970090.2 | XP_012825544.1 | 3 - 4 | 2715 | 75 | ||
XM_012970091.2 | XP_012825545.1 | 3 - 4 | 340 | 75 | ||
XM_012970092.2 | XP_012825546.1 | 3 - 4 | 219 | 75 | ||
XM_018093785.1 | XP_017949274.1 | 3 - 4 | 326 | 75 | ||
GenBank mRNA | BC161143.1 | - | 3 - 4 | 322 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |