Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankdd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127008.1 | NP_001120480.1 | 5 - 6 | 697 | 122 | |
XM_012956326.2 | XP_012811780.1 | 5 - 6 | 545 | 122 | ||
XM_018091063.1 | XP_017946552.1 | 5 - 6 | 569 | 122 | ||
GenBank mRNA | BC161348.1 | - | 5 - 6 | 697 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |