Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127071.1 | NP_001120543.1 | 4 - 5 | 659 | 78 | |
XM_012960633.2 | XP_012816087.1 | 3 - 4 | 870 | 78 | ||
XM_012960634.2 | XP_012816088.1 | 5 - 6 | 1438 | 78 | ||
GenBank mRNA | BC161470.1 | - | 4 - 5 | 659 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |