Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127130.1 | NP_001120602.1 | 11 - 12 | 1363 | 98 | |
GenBank mRNA | BC161637.1 | - | 11 - 12 | 1363 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |