Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114204.1 | NP_001107676.1 | 3 - 4 | 583 | 112 | |
GenBank mRNA | BC087818.1 | - | 3 - 4 | 519 | 112 | |
BC157309.1 | - | 3 - 4 | 583 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |