Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126600.1 | NP_001120072.1 | 1 - 2 | 419 | 95 | |
GenBank mRNA | BC158495.1 | - | 1 - 2 | 419 | 95 | |
CR848061.2 | - | 1 - 2 | 310 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |