Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126759.1 | NP_001120231.1 | 5 - 6 | 421 | 87 | |
GenBank mRNA | BC160408.1 | - | 5 - 6 | 421 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |