Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126762.1 | NP_001120234.1 | 2 - 3 | 302 | 98 | |
XM_012962724.2 | XP_012818178.2 | 2 - 3 | 381 | 98 | ||
XM_012962725.2 | XP_012818179.1 | 2 - 3 | 337 | 98 | ||
XM_012962726.2 | XP_012818180.1 | 2 - 3 | 296 | 98 | ||
XM_012962727.2 | XP_012818181.1 | 2 - 3 | 280 | 98 | ||
GenBank mRNA | BC160413.1 | - | 2 - 3 | 302 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |