Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126905.1 | NP_001120377.1 | 10 - 11 | 1350 | 98 | |
GenBank mRNA | BC161074.1 | - | 10 - 11 | 1350 | 98 | |
CT030494.1 | - | 7 - 8 | 1118 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |