Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpatch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126999.1 | NP_001120471.1 | 1 - 2 | 330 | 88 | |
GenBank mRNA | BC161307.1 | - | 1 - 2 | 330 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |