Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hypk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127003.1 | NP_001120475.1 | 1 - 2 | 168 | 100 | |
GenBank mRNA | BC161322.1 | - | 1 - 2 | 168 | 100 | |
CR942732.2 | - | 1 - 2 | 165 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |