Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: maats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127421.1 | NP_001120893.1 | 16 - 17 | 2146 | 67 | |
GenBank mRNA | BC166178.1 | - | 16 - 17 | 2146 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |