Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siglec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127949.1 | NP_001121421.1 | 949 | 104 | ||
GenBank mRNA | BC166151.1 | - | 949 | 104 |
Gene Symbol: LOC101732774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018095750.1 | XP_017951239.1 | 3 - 4 | 432 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |