Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128005.1 | NP_001121477.1 | 2 - 3 | 653 | 96 | |
GenBank mRNA | BC166250.1 | - | 2 - 3 | 653 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |