Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rspo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128028.1 | NP_001121500.1 | 2 - 3 | 647 | 70 | |
GenBank mRNA | BC166324.1 | - | 2 - 3 | 647 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |