Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128640.1 | NP_001122112.1 | 4 - 5 | 2892 | 84 | |
XM_012964011.2 | XP_012819465.1 | 3 - 4 | 3117 | 84 | ||
XM_012964013.2 | XP_012819467.1 | 3 - 4 | 3158 | 84 | ||
XM_012964015.2 | XP_012819469.1 | 2 - 3 | 2896 | 84 | ||
XM_012964018.2 | XP_012819472.1 | 2 - 3 | 3022 | 84 | ||
GenBank mRNA | BC167678.1 | - | 4 - 5 | 2892 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |