Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129936.1 | NP_001123408.1 | 4 - 5 | 1368 | 83 | |
GenBank mRNA | BC167403.1 | - | 4 - 5 | 1368 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |