Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130237.1 | NP_001123709.1 | 2 - 3 | 217 | 102 | |
XM_012958647.2 | XP_012814101.1 | 3 - 4 | 406 | 102 | ||
XM_018091950.1 | XP_017947439.1 | 3 - 4 | 229 | 102 | ||
GenBank mRNA | BC167315.1 | - | 2 - 3 | 217 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |