Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130237.1 | NP_001123709.1 | 3 - 4 | 255 | 91 | |
XM_012958647.2 | XP_012814101.1 | 4 - 5 | 444 | 91 | ||
XM_018091950.1 | XP_017947439.1 | 4 - 5 | 267 | 91 | ||
GenBank mRNA | BC167315.1 | - | 3 - 4 | 255 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |