Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ildr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130373.1 | NP_001123845.1 | 3 - 4 | 639 | 89 | |
GenBank mRNA | BC167711.1 | - | 3 - 4 | 639 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |