Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142044.1 | NP_001135516.1 | 2 - 3 | 644 | 89 | |
XM_012956554.1 | XP_012812008.1 | 3 - 4 | 557 | 89 | ||
XM_012956555.1 | XP_012812009.1 | 3 - 4 | 557 | 89 | ||
XM_012956556.1 | XP_012812010.1 | 2 - 3 | 694 | 89 | ||
XM_012956557.1 | XP_012812011.1 | 2 - 3 | 322 | 89 | ||
GenBank mRNA | BC167935.2 | - | 2 - 3 | 644 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |