Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142068.1 | NP_001135540.1 | 7 - 8 | 1552 | 73 | |
XM_012966199.2 | XP_012821653.1 | 8 - 9 | 1894 | 73 | ||
XM_012966200.2 | XP_012821654.1 | 8 - 9 | 1892 | 73 | ||
XM_012966201.2 | XP_012821655.1 | 8 - 9 | 1883 | 73 | ||
XM_012966202.2 | XP_012821656.1 | 8 - 9 | 1880 | 73 | ||
XM_012966203.2 | XP_012821657.1 | 7 - 8 | 1822 | 73 | ||
XM_012966204.2 | XP_012821658.1 | 7 - 8 | 1810 | 73 | ||
XM_012966206.2 | XP_012821660.1 | 8 - 9 | 1893 | 73 | ||
XM_012966207.2 | XP_012821661.1 | 8 - 9 | 1890 | 73 | ||
XM_012966208.2 | XP_012821662.1 | 7 - 8 | 1820 | 73 | ||
XM_012966209.2 | XP_012821663.1 | 7 - 8 | 1817 | 73 | ||
XM_012966210.2 | XP_012821664.1 | 7 - 8 | 1809 | 73 | ||
XM_018095295.1 | XP_017950784.1 | 8 - 9 | 1894 | 73 | ||
GenBank mRNA | BC168010.1 | - | 7 - 8 | 1552 | 73 | |
CT030480.1 | - | 5 - 6 | 1207 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |