Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142137.1 | NP_001135609.1 | 3 - 4 | 668 | 90 | |
XM_012956572.2 | XP_012812026.1 | 3 - 4 | 873 | 90 | ||
XM_012956573.2 | XP_012812027.1 | 3 - 4 | 667 | 90 | ||
XM_018091112.1 | XP_017946601.1 | 3 - 4 | 885 | 90 | ||
GenBank mRNA | BC168429.1 | - | 3 - 4 | 668 | 90 | |
CR942589.2 | - | 3 - 4 | 521 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |