Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142210.1 | NP_001135682.1 | 5 - 6 | 475 | 74 | |
XM_012966233.2 | XP_012821687.1 | 5 - 6 | 805 | 74 | ||
XM_012966234.2 | XP_012821688.1 | 5 - 6 | 683 | 74 | ||
XM_012966235.2 | XP_012821689.1 | 5 - 6 | 688 | 74 | ||
GenBank mRNA | BC168609.1 | - | 5 - 6 | 475 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |