Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142234.1 | NP_001135706.1 | 4 - 5 | 891 | 75 | |
XM_012957657.1 | XP_012813111.1 | 5 - 6 | 814 | 75 | ||
GenBank mRNA | BC168820.1 | - | 4 - 5 | 891 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |