Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142241.1 | NP_001135713.1 | 6 - 7 | 2400 | 70 | |
XM_012956584.2 | XP_012812038.1 | 7 - 8 | 1171 | 70 | ||
GenBank mRNA | BC169148.1 | - | 6 - 7 | 2400 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |