Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142250.1 | NP_001135722.1 | 2 - 3 | 217 | 89 | |
GenBank mRNA | BC169180.1 | - | 2 - 3 | 217 | 89 | |
CU075403.1 | - | 2 - 3 | 205 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |