Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142826.2 | NP_001136298.1 | 24 - 25 | 1038 | 105 | |
XM_012970194.2 | XP_012825648.1 | 23 - 24 | 1016 | 105 | ||
XM_012970195.2 | XP_012825649.1 | 24 - 25 | 1058 | 105 | ||
GenBank mRNA | CU075851.1 | - | 24 - 25 | 1191 | 105 | |
EU220008.1 | - | 24 - 25 | 1040 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |