Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142826.2 | NP_001136298.1 | 5 - 6 | 296 | 90 | |
XM_012970193.2 | XP_012825647.1 | 5 - 6 | 316 | 90 | ||
XM_012970194.2 | XP_012825648.1 | 5 - 6 | 316 | 90 | ||
XM_012970195.2 | XP_012825649.1 | 5 - 6 | 316 | 90 | ||
XM_012970196.2 | XP_012825650.1 | 5 - 6 | 316 | 90 | ||
XM_012970197.2 | XP_012825651.1 | 5 - 6 | 316 | 90 | ||
GenBank mRNA | CU075851.1 | - | 5 - 6 | 449 | 90 | |
EU220008.1 | - | 5 - 6 | 298 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |