Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145376.2 | NP_001138848.1 | 3 - 4 | 346 | 97 | |
GenBank mRNA | BC080491.1 | - | 3 - 4 | 348 | 97 | |
BC158910.1 | - | 3 - 4 | 310 | 97 | ||
CR760967.2 | - | 3 - 4 | 283 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |