Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145376.2 | NP_001138848.1 | 4 - 5 | 511 | 100 | |
GenBank mRNA | BC080491.1 | - | 4 - 5 | 513 | 100 | |
BC158910.1 | - | 4 - 5 | 475 | 100 | ||
CR760967.2 | - | 4 - 5 | 448 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |