Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145376.2 | NP_001138848.1 | 8 - 9 | 1152 | 78 | |
GenBank mRNA | BC080491.1 | - | 8 - 9 | 1154 | 78 | |
BC158910.1 | - | 8 - 9 | 1116 | 78 | ||
CR760967.2 | - | 8 - 9 | 1089 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |