Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016735.2 | NP_001016735.1 | 5 | 1852 | 100 | |
GenBank mRNA | CR760014.2 | - | 5 | 1852 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |