Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102986.1 | NP_001096456.1 | 1 | 954 | 108 | |
XM_012956312.1 | XP_012811766.1 | 2 | 891 | 108 | ||
GenBank mRNA | BC124005.1 | - | 1 | 905 | 108 | |
BC136178.1 | - | 1 | 954 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |